Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ 10X RBC Lysis Buffer (Multi-species)

Catalog No. 501129743
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9743 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-9743 Supplier Invitrogen™ Supplier No. 00430054
  • $96.00
    $71.65 / Each of 1

    Save $24.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Specially formulated for optimal lysis of erythrocytes in single-cell suspensions of peripheral blood and hematopoietic tissues such as spleen. This buffer can be used for lysis of human, mouse, rat, canine, and non-human primate samples.

Contains ammonium chloride, which lyses red blood cells with a minimal effect on lymphocytes when used as instructed.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Format Liquid
Product Type Lysis Buffer
Volume (Metric) 50 mL
Product Line eBioscience
We've purchased the eBioscience 10X RBC Lysis Buffer (Multi-species) (Cat. No. 00-4300-54). Can we store the reconstituted solution at room temp or is it necessary to prepare the solution fresh before use?

The diluted 10X RBC Lysis Buffer should be stable for about 1 month when stored at room temperature. The product will be more stable as a 10X concentrate. We recommend making enough of the 1X solution to last 3-4 weeks.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.