Learn More
FUJIFILM Biosciences rBC2 LCN 635 AiLecS1 100UL
Catalog No. 5000450329
Encompass_Preferred
rBC2LCN-635 AiLecS1-635 is a recombinant lectin conjugated with a 635 dye designed for sensitive and specific fluorescent labeling of glycan structures on cell surfaces Ideal for cell imaging and analysis with minimal background signal This is a recombinant lectin rBC2LCN that can specifically recognize undifferentiated cells and has already been labeled with fluorescent dye 635 rBC2LCN specifically recognize a sugar chain which exists on the surface of hPSCs When you add rBC2LCN-635 to the medium which is used for hPSCs culture you can stain the living hPSCs eBC2LCN-635 is not toxic to cells therefore you can culture hPSCs without any effect on the growth and pluripotency in the medium containing rBC2LCN-635 This product can be used for both cell staining and flow cytometry Excitation 634 nm Emission 654 nm
Catalog No. 50-004-50329 Supplier FUJIFILM Biosciences Supplier No. 18503161
rBC2LCN-635 AiLecS1-635 is a recombinant lectin conjugated with a 635 dye designed for sensitive and specific fluorescent labeling of glycan structures on cell surfaces Ideal for cell imaging and analysis with minimal background signal This is a recombinant lectin rBC2LCN that can specifically recognize undifferentiated cells and has already been labeled with fluorescent dye 635 rBC2LCN specifically recognize a sugar chain which exists on the surface of hPSCs When you add rBC2LCN-635 to the medium which is used for hPSCs culture you can stain the living hPSCs eBC2LCN-635 is not toxic to cells therefore you can culture hPSCs without any effect on the growth and pluripotency in the medium containing rBC2LCN-635 This product can be used for both cell staining and flow cytometry Excitation 634 nm Emission 654 nm