Learn More
Eurofins Discoverx RBL-2H3 ADORA2A Gq Cell Line, 2 vials. (95-1008C11)
Catalog No. 502004055
Encompass
The ADORA2A Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. adenosine, NECA) based activation of ADORA2A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADORA2A, a naturally Gq coupled GPCR. Upon activation of ADORA2A, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4055 Supplier Eurofins Discoverx Supplier No. 951008C11
Add to Cart
Add to Cart
Request a Quote
The ADORA2A Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. adenosine, NECA) based activation of ADORA2A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADORA2A, a naturally Gq coupled GPCR. Upon activation of ADORA2A, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.