Learn More
Aviva Systems Biology Corp RBM10 Antibody - N-terminal region (ARP30103_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30103_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RBM10. Alternate names: S1-1, TARPS, GPATC9, ZRANB5, GPATCH9, DXS8237EApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54124