Learn More
Aviva Systems Biology Corp RBM12 Antibody - N-terminal region (ARP40623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBM12. Alternate names: HRIHFB2091, KIAA0765, SWANApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57553