Learn More
Aviva Systems Biology Corp RBM18 Antibody - middle region (ARP41071_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41071_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM18. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RBM18. Alternate names: MGC2734ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-910