Learn More
Aviva Systems Biology Corp RBM22 Antibody - middle region (ARP40888_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40888_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM22. Alternate names: FLJ10290, ZC3H16, Cwc2, fSAP47ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57653