Learn More
Aviva Systems Biology Corp RBM22 Antibody - C-terminal region (ARP40889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM22. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RBM22. Alternate names: FLJ10290, Cwc2, ZC3H16, fSAP47ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57654