Learn More
Aviva Systems Biology Corp Rbm25 Antibody - middle region (ARP40968_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40968_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rbm25. It was validated on Western Blot. . Alternate names: 2600011C06Rik, 2610015J01Rik, A130095G20Rik, AI159652, AL023075, AU043498, RNPC7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-906