Learn More
Aviva Systems Biology Corp RBM26 Antibody - middle region (ARP40975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM26. Alternate names: ARRS2, SE70-2, ZC3H17, PRO1777, C13orf10, RP11-255E21.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57682