Learn More
Aviva Systems Biology Corp Rbm3 Antibody - N-terminal region (ARP40307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rbm3. It was validated on Western Blot. . Alternate names: MGC105811ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-284-864