Learn More
Aviva Systems Biology Corp RBM35A Antibody - N-terminal region (ARP42489_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42489_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM35A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBM35A. Alternate names: FLJ20171, RBM35A, RMB35AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58245