Learn More
Aviva Systems Biology Corp RBM35B Antibody - N-terminal region (ARP41016_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41016_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM35B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBM35B. Alternate names: FLJ21918, FLJ22248, RBM35BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53260