Learn More
Aviva Systems Biology Corp RBM38 Antibody - N-terminal region (ARP40862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM38. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RBM38. Alternate names: RNPC1, SEB4B, SEB4D, HSRNASEB, dJ800J21.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53246