Learn More
Aviva Systems Biology Corp RBM38 Antibody - middle region (ARP40863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM38. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM38. Alternate names: HSRNASEB, RNPC1, SEB4B, SEB4D, dJ800J21.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57643