Learn More
Aviva Systems Biology Corp RBM39 Antibody - N-terminal region (ARP40560_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40560_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBM39. Alternate names: CAPER, CAPERalpha, CC1.3, DKFZp781C0423, FLJ44170, HCC1, RNPC2, FSAP59ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53206