Learn More
Aviva Systems Biology Corp RBM4 Antibody - middle region (ARP40426_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40426_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM4. Alternate names: DKFZp547K0918, LARK, MGC75138, RBM4A, ZCCHC21, ZCRB3AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57443