Learn More
Aviva Systems Biology Corp RBM4 Antibody - C-terminal region (ARP40425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RBM4. Alternate names: DKFZp547K0918, LARK, MGC75138, RBM4A, ZCCHC21, ZCRB3A, hLARKApplicationWB. Species reactivity: Human
Catalog No. 50-014-53197