Learn More
Aviva Systems Biology Corp RBM45 Antibody - middle region (ARP41154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM45. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RBM45. Alternate names: DRB1, FLJ44612ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53282