Learn More
Aviva Systems Biology Corp RBM47 Antibody - middle region (ARP40932_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40932_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM47. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM47. Alternate names: DKFZp686F02235, FLJ20273, FLJ21344, FLJ21643, NET18ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57668