Learn More
Aviva Systems Biology Corp RBM4B Antibody - C-terminal region (ARP41039_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41039_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM4B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RBM4B. Alternate names: RBM30, RBM4L, ZCRB3B, ZCCHC15ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57700