Learn More
Aviva Systems Biology Corp RBM6 Antibody - N-terminal region (ARP31461_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31461_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBM6. Alternate names: 3G2, DEF-3, DEF3, DKFZp686B0877, FLJ36517, HLC-11, NY-LU-12, g16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54234