Learn More
Aviva Systems Biology Corp RBM7 Antibody - middle region (ARP40830_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40830_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM7. Alternate names: FLJ11153ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57633