Learn More
Aviva Systems Biology Corp RBM8A Antibody - N-terminal region (ARP40576_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40576_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM8A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RBM8A. Alternate names: Y14, RBM8, ZNRP, RBM8B, ZRNP1, BOV-1A, BOV-1B, BOV-1C, MDS014ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53208