Learn More
Aviva Systems Biology Corp RBM9 Antibody - N-terminal region (ARP40340_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40340_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBM9. Alternate names: RTA, fxh, FOX2, RBM9, Fox-2, HNRBP2, HRNBP2, dJ106I20.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57398