Learn More
Aviva Systems Biology Corp Rbm9 Antibody - C-terminal region (ARP37433_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37433_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rbm9. It was validated on Western Blot. Rbfox2,RNA binding protein,.x-1 homolog (C. elegans) 2,Fxh; Fbm2; Rbm9; Hrnbp2; AA407676; AI118529; 2810460A15Rik; Rbfox2,RNA-binding protein 9,RNA-binding protein 9,fox-1 homolog B,fox-1 homolog Fxh,Fyn-binding molecule 2,RNA binding motif protein 9,RNA-binding motif protein 9,hexaribonucleotide binding protein 2,hexaribonucleotide-binding protein 2The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2810460A15Rik, AA407676, AI118529, Fbm2, Fxh, Hrnbp2, Rbm9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-631