Learn More
Aviva Systems Biology Corp RBM9 Antibody - middle region (ARP40341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBM9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBM9. Alternate names: Fox-2, HNRBP2, HRNBP2, RTA, dJ106I20.3, fxh, FOX2, RBM9ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57399