Learn More
Aviva Systems Biology Corp RBMS1 Antibody - C-terminal region (ARP40427_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40427_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBMS1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RBMS1. Alternate names: YC1, MSSP, SCR2, HCC-4, MSSP-1, MSSP-2, MSSP-3, C2orf12ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57444