Learn More
Aviva Systems Biology Corp RBMS3 Antibody - N-terminal region (ARP40234_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40234_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBMS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBMS3. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57358