Learn More
Aviva Systems Biology Corp RBMX Antibody - C-terminal region (ARP61802_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61802_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBMX. It was validated on Western Blot. . Alternate names: HNRPG, RBMXP1, RBMXRT, RNMX, hnRNP-GApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-472