Learn More
Aviva Systems Biology Corp RBMY1A1 Antibody - N-terminal region (ARP40257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBMY1A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBMY1A1. Alternate names: RBM1, RBM2, RBMY, YRRM1, YRRM2, RBM, RBMY1CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53183