Learn More
AdipoGen RBN-2397
SDP
Catalog No. 502562305
Encompass_Preferred
Chemical. CAS 2381037-82-5. Formula C20H23F6N7O3. MW 523.4. RBN-2397 is a potent NAD competitive small molecule inhibitor of PARP-7. RBN-2397 selectively inhibits PARP-7 and demonstrates >50-fold selectivity for inhibition of PARP-7 over all PARP family members as measured by biochemical assays. RBN-2397 inhibits PARP-7-dependent MARylation IC50 = 2nM in cell biochemical assay and inhibits cells proliferation in NCI-H1373 lung cancer cells IC50 = 20nM. By selectively inhibiting PARP-7 in tumor cells, RBN-2397 has been shown to directly inhibit cellular proliferation and restore Type I interferon signaling to stimulate an innate and adaptive antitumor immune response. Targeting cytosolic nucleic acid sensing pathways and the Type I interferon IFN response is an emerging therapeutic strategy being explored in oncology. The PARP family consists of seventeen enzymes that regulate fundamental biological processes including response to cellular stress.
Catalog No. 50-256-2305 Supplier AdipoGen Supplier No. AGCR13547M025
Chemical. CAS 2381037-82-5. Formula C20H23F6N7O3. MW 523.4. RBN-2397 is a potent NAD competitive small molecule inhibitor of PARP-7. RBN-2397 selectively inhibits PARP-7 and demonstrates >50-fold selectivity for inhibition of PARP-7 over all PARP family members as measured by biochemical assays. RBN-2397 inhibits PARP-7-dependent MARylation IC50 = 2nM in cell biochemical assay and inhibits cells proliferation in NCI-H1373 lung cancer cells IC50 = 20nM. By selectively inhibiting PARP-7 in tumor cells, RBN-2397 has been shown to directly inhibit cellular proliferation and restore Type I interferon signaling to stimulate an innate and adaptive antitumor immune response. Targeting cytosolic nucleic acid sensing pathways and the Type I interferon IFN response is an emerging therapeutic strategy being explored in oncology. The PARP family consists of seventeen enzymes that regulate fundamental biological processes including response to cellular stress.