Learn More
TARGETMOL CHEMICALS INC RBN-2397 5MG
SDP
Catalog No. 502581431
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RBN-2397 is a potent selective and orally active accross species NAD+ competitive PARP7 inhibitor with IC50 less than 3 nM. purity: 99%
Catalog No. 50-258-1431 Supplier TARGETMOL CHEMICALS INC Supplier No. T126955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RBN-2397 is a potent selective and orally active accross species NAD+ competitive PARP7 inhibitor with IC50 less than 3 nM. purity: 99%