Learn More
Medchemexpress LLC Rbn-3143 | 2360853-16-1 | 99.3% | 449.54 g/mol | C22H28FN3O4S | 10 MG
SDP
Catalog No. 5000286723
Encompass_Preferred
RBN-3143 is a small-molecule, NAD+-competitive catalytic inhibitor of PARP14 with reported nanomolar potency (IC50 ≈ 4 nM). It inhibits PARP14-mediated ADP-ribosylation, stabilizes PARP14 in cell lines, and is provided for research use to support studies of PARP14 biology and inflammatory disease models.
  • NAD+-competitive catalytic inhibitor of PARP14.
  • Reported IC50 ≈ 4 nM against PARP14.
  • Inhibits PARP14-mediated ADP-ribosylation in cell assays.
  • Stabilizes PARP14 protein in cell lines.
  • High reported chemical purity (~99.3%).
  • Available in multiple research pack sizes for laboratory studies.
Catalog No. 50-002-86723 Supplier Medchemexpress LLC Supplier No. HY15020710MG
RBN-3143 is a small-molecule, NAD+-competitive catalytic inhibitor of PARP14 with reported nanomolar potency (IC50 ≈ 4 nM). It inhibits PARP14-mediated ADP-ribosylation, stabilizes PARP14 in cell lines, and is provided for research use to support studies of PARP14 biology and inflammatory disease models.
  • NAD+-competitive catalytic inhibitor of PARP14.
  • Reported IC50 ≈ 4 nM against PARP14.
  • Inhibits PARP14-mediated ADP-ribosylation in cell assays.
  • Stabilizes PARP14 protein in cell lines.
  • High reported chemical purity (~99.3%).
  • Available in multiple research pack sizes for laboratory studies.