Learn More
Medchemexpress LLC RBN-3143 | 2360853-16-1 | C22H28FN3O4S | 1 ML
SDP
Catalog No. 5000375447
Encompass_Preferred
RBN-3143 is a potent and NAD+-competitive catalytic PARP14 inhibitor with an IC50 value of 4 nM. It inhibits PARP14-mediated ADP-ribosylation and stabilizes PARP14 in cell lines. RBN-3143 can be used in research of lung inflammation.
  • Potent and NAD+-competitive catalytic PARP14 inhibitor.
  • Inhibits PARP14-mediated ADP-ribosylation and stabilizes PARP14 in cell lines.
  • Can be used in research of lung inflammation.
  • In vivo, RBN-3143 (5 μg; intranasal administration) can improve steroid-resistant asthma mouse models.
  • Suppressed the accumulation of alarmins TSLP, IL-33, and IL-25.
Catalog No. 50-003-75447 Supplier Medchemexpress LLC Supplier No. HY15020710MM/1ML
Add to Cart
Add to Cart
RBN-3143 is a potent and NAD+-competitive catalytic PARP14 inhibitor with an IC50 value of 4 nM. It inhibits PARP14-mediated ADP-ribosylation and stabilizes PARP14 in cell lines. RBN-3143 can be used in research of lung inflammation.
  • Potent and NAD+-competitive catalytic PARP14 inhibitor.
  • Inhibits PARP14-mediated ADP-ribosylation and stabilizes PARP14 in cell lines.
  • Can be used in research of lung inflammation.
  • In vivo, RBN-3143 (5 μg; intranasal administration) can improve steroid-resistant asthma mouse models.
  • Suppressed the accumulation of alarmins TSLP, IL-33, and IL-25.