Learn More
Medchemexpress LLC RBN-3143 | 2360853-16-1 | C22H28FN3O4S | 100 MG
SDP
Catalog No. 5000374208
Encompass_Preferred
RBN-3143 is a potent and selective inhibitor targeting PARP14, exhibiting an IC50 value of 4 nM. This compound effectively inhibits PARP14-mediated ADP-ribosylation and helps stabilize PARP14 in cellular models. It has shown promise in preclinical studies related to lung inflammation.
  • Potent and NAD+-competitive PARP14 inhibitor.
  • Demonstrates an IC50 of 4 nM against PARP14.
  • Inhibits ADP-ribosylation activity of PARP14.
  • Stabilizes PARP14 in cell lines.
  • Investigated for its potential in lung inflammation research.
Catalog No. 50-003-74208 Supplier Medchemexpress LLC Supplier No. HY150207100MG
Add to Cart
Add to Cart
RBN-3143 is a potent and selective inhibitor targeting PARP14, exhibiting an IC50 value of 4 nM. This compound effectively inhibits PARP14-mediated ADP-ribosylation and helps stabilize PARP14 in cellular models. It has shown promise in preclinical studies related to lung inflammation.
  • Potent and NAD+-competitive PARP14 inhibitor.
  • Demonstrates an IC50 of 4 nM against PARP14.
  • Inhibits ADP-ribosylation activity of PARP14.
  • Stabilizes PARP14 in cell lines.
  • Investigated for its potential in lung inflammation research.