Learn More
Medchemexpress LLC RBN-3143 | 2360853-16-1 | C22H28FN3O4S | 5 MG
SDP
Catalog No. 5000375692
Encompass_Preferred
RBN-3143 is a potent and NAD+-competitive catalytic PARP14 inhibitor with an IC50 value of 4 nM. It inhibits PARP14-mediated ADP-ribosylation and stabilizes PARP14 in cell lines. It can be used in research of lung inflammation.
  • Potent and NAD+-competitive catalytic PARP14 inhibitor
  • Inhibits PARP14-mediated ADP-ribosylation
  • Stabilizes PARP14 in cell lines
  • Used in research of lung inflammation
  • Improves steroid-resistant asthma mouse models
  • Suppresses alarmins TSLP, IL-33, and IL-25 in mouse models of asthma
Catalog No. 50-003-75692 Supplier Medchemexpress LLC Supplier No. HY1502075MG
Add to Cart
Add to Cart
RBN-3143 is a potent and NAD+-competitive catalytic PARP14 inhibitor with an IC50 value of 4 nM. It inhibits PARP14-mediated ADP-ribosylation and stabilizes PARP14 in cell lines. It can be used in research of lung inflammation.
  • Potent and NAD+-competitive catalytic PARP14 inhibitor
  • Inhibits PARP14-mediated ADP-ribosylation
  • Stabilizes PARP14 in cell lines
  • Used in research of lung inflammation
  • Improves steroid-resistant asthma mouse models
  • Suppresses alarmins TSLP, IL-33, and IL-25 in mouse models of asthma