Learn More
Medchemexpress LLC HY-136979 5mg Medchemexpress, RBN012759 CAS:2360851-29-0 Purity:>98%
SDP
Catalog No. 502030469
Encompass_Preferred
Medchemexpress, HY-136979 5mg RBN012759 CAS:2360851-29-0 RBN012759 is a potent, selective and orally active inhibitor of PARP14, with an IC50 of <3 nM. RBN012759 displays 300-fold selectivity over the monoPARPs and 1000-fold selectivity over the polyPARPs. RBN012759 decreases pro-tumor macrophage function and elicits inflammatory responses in tumor explants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0469 Supplier Medchemexpress LLC Supplier No. HY1369795MG
Medchemexpress, HY-136979 5mg RBN012759 CAS:2360851-29-0 RBN012759 is a potent, selective and orally active inhibitor of PARP14, with an IC50 of <3 nM. RBN012759 displays 300-fold selectivity over the monoPARPs and 1000-fold selectivity over the polyPARPs. RBN012759 decreases pro-tumor macrophage function and elicits inflammatory responses in tumor explants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.