Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5947MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IF, IHC. Conjugate/Tag: Unconjugated. Clone: G4E4. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Hepatic or ovarian carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Retinol binding protein-1 purified from human plasma. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 21-25kDa.Chromosome: 10q23.33. Gene Symbol: RBP1. Gene ID: 5950. Ensemble: ENSG00000138207. Uniprot: NP_006735.
Catalog No. 50-188-5127