Learn More
Aviva Systems Biology Corp RBP1 Antibody - middle region (ARP42081_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42081_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBP1. Alternate names: CRABP-I, CRBP, CRBP1, RBPC, CRBPIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58100