Learn More
Aviva Systems Biology Corp RBP4 Antibody - N-terminal region (ARP41578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBP4. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-57913