Learn More
Genscript Corporation RBP4 His Human
Supplier:  Genscript Corporation Z0337850
Encompass
The properties of retinol binding protein is the transport carrier of vitamin A in the plasma Human-retinol binding protein is a single-chain polypeptide with a molecular weight of approximately 21000 and one binding site for retinol and other forms of vitamin A In addition compounds related to retinol such as retinal retinoic acid retinyl esters and geometric isomers of retinol and of retinal were evaluated for their ability to bind to this protein In plasma RBP4-retinol forms a complex with transthyretin (TTR) also known as thyroxine-binding protein and prealbumin Defects in RBP4 cause retinol-binding protein deficiency which affects night vision
Catalog No. 50-000-56992