Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RBP4 Human HEK293 100ug
SDP

Catalog No. 5000195498
Encompass_Preferred

Recombinant human retinol-binding protein 4 (RBP4) expressed in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized and purified to >95% by reducing SDS-PAGE, this protein is provided in multiple pack sizes, including a 100 μg option, for research applications. Formulated in 20 mM phosphate buffer, 150 mM NaCl, pH 7.8; store at -20°C and aliquot for long-term storage at -80°C.

  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation for improved stability and convenient storage.
  • Available in multiple sizes, including a 100 μg pack.
  • Contains amino acids E19-L201; approximate molecular weight ~21 kDa.
  • Intended for research use only.

Catalog No. 50-001-95498 Supplier Medchemexpress LLC Supplier No. HYP72031A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human retinol-binding protein 4 (RBP4) expressed in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized and purified to >95% by reducing SDS-PAGE, this protein is provided in multiple pack sizes, including a 100 μg option, for research applications. Formulated in 20 mM phosphate buffer, 150 mM NaCl, pH 7.8; store at -20°C and aliquot for long-term storage at -80°C.

  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation for improved stability and convenient storage.
  • Available in multiple sizes, including a 100 μg pack.
  • Contains amino acids E19-L201; approximate molecular weight ~21 kDa.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.