Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RBP4 Human HEK293 100ug
SDP

Supplier:  Medchemexpress LLC HYP72031100UG

Encompass_Preferred

Recombinant human Retinol Binding Protein 4 (RBP4) produced in HEK293 cells with a C-terminal human Fc tag, supplied for research use in biochemical and cell-based assays. Available in common research-scale quantities including a 100 μg format, the protein is purified to high purity and supplied with storage guidance to preserve activity.

  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal human Fc tag facilitates detection and purification.
  • Available in small research-scale quantities, including 100 μg.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 53 kDa.
  • Recommended storage at -20°C or -80°C; aliquot to avoid freeze-thaw.

Catalog No. 50-001-95499


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.