Learn More
Medchemexpress LLC RBP4 Human HEK293 50ug
SDP
Catalog No. 5000195497
Encompass_Preferred
Recombinant human retinol-binding protein 4 (RBP4) produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a 50 μg vial for research use, this purified protein is suitable for biochemical assays, antibody validation, and immunodetection applications; follow the product datasheet for storage and handling recommendations.
  • Expressed in HEK293 cells to preserve human-like post-translational modifications.
  • C-terminal 6xHis tag facilitates detection and purification.
  • Purified recombinant protein suitable for biochemical assays and antibody validation.
  • Supplied as a 50 μg vial for small-scale experiments; store and handle per recommendations.
Catalog No. 50-001-95497 Supplier Medchemexpress LLC Supplier No. HYP72031A50UG
Recombinant human retinol-binding protein 4 (RBP4) produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a 50 μg vial for research use, this purified protein is suitable for biochemical assays, antibody validation, and immunodetection applications; follow the product datasheet for storage and handling recommendations.
  • Expressed in HEK293 cells to preserve human-like post-translational modifications.
  • C-terminal 6xHis tag facilitates detection and purification.
  • Purified recombinant protein suitable for biochemical assays and antibody validation.
  • Supplied as a 50 μg vial for small-scale experiments; store and handle per recommendations.