Learn More
AdipoGen RBP4 human pAb
SDP
Supplier:  AdipoGen AG25A0053C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human RBP4. Detects a band of ~22-25kDa by Western blot. Weakly cross-reacts with mouse and rat RBP4. Source Guinea pig. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Retinol binding protein 4 RBP4 RBP is a 21kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol vitamin A to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 GLUT4 suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity, and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function.
Catalog No. 50-256-3259