Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RBP4 inhibitor 1 | 1198180-03-8 | 99.7% | 357.25 | C14H13F6NO3 | 5 MG
SDP

Catalog No. 5000222771
Encompass_Preferred

RBP4 inhibitor 1 is a potent, orally active retinol-binding protein 4 (RBP4) inhibitor reported to reduce blood RBP4 levels in vivo. It exhibits low-nanomolar activity against human RBP4 and is supplied as a high-purity research compound.

  • Potent RBP4 inhibition: IC50 28 nM (human), 110 nM (mouse).
  • High purity: 99.74%.
  • Appearance: white to off-white solid.
  • Molecular weight: 357.25 g/mol.
  • Chemical formula: C14H13F6NO3.
  • SMILES string provided for structural reference.
  • Storage: powder at -20°C (stable for 3 years); in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in small research pack sizes and DMSO solutions for screening.

Catalog No. 50-002-22771 Supplier Medchemexpress LLC Supplier No. HY1437935MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RBP4 inhibitor 1 is a potent, orally active retinol-binding protein 4 (RBP4) inhibitor reported to reduce blood RBP4 levels in vivo. It exhibits low-nanomolar activity against human RBP4 and is supplied as a high-purity research compound.

  • Potent RBP4 inhibition: IC50 28 nM (human), 110 nM (mouse).
  • High purity: 99.74%.
  • Appearance: white to off-white solid.
  • Molecular weight: 357.25 g/mol.
  • Chemical formula: C14H13F6NO3.
  • SMILES string provided for structural reference.
  • Storage: powder at -20°C (stable for 3 years); in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in small research pack sizes and DMSO solutions for screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.