Learn More
Proteintech Group Inc RBP7 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 145411AP150UL
Encompass_Preferred
The RBP7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RBP7. This antibody recognizes human, mouse, rat antigen. The RBP7 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Catalog No. 50-225-6394