Learn More
ABclonal Technology RBP7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A159391000UL
Encompass_Preferred
The protein encoded by this gene is a member of the cellular retinol-binding protein (CRBP) family, whose members are required for vitamin A stability and metabolism. The encoded protein binds all-trans-retinol and is structurally similar to other CRBPs, however, it has a lower binding affinity for retinol than other CRBPs.
Catalog No. 50-001-07120