Learn More
Medchemexpress LLC HY-137471 5mg Medchemexpress, RBPJ Inhibitor-1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1374715MG
Encompass_Preferred
Medchemexpress, HY-137471 5mg RBPJ Inhibitor-1 CAS: RBPJ Inhibitor-1 (RIN1), the first RBPJ inhibitor, blocks the functional interaction of RBPJ with SHARP. RBPJ Inhibitor-1 (RIN1) inhibits NOTCH-dependent tumor cell proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1490